Learn More
DCP1B Rabbit anti-Human, Polyclonal, Bethyl Laboratories
Rabbit Polyclonal Antibody
Supplier: Bethyl Laboratories, Inc A303641A

Description
The recommended shelf life for this product is 1 year from date of receipt. Based on 100% sequence identity, this antibody is predicted to react with Orangutan
DCP1B is a core component of the mRNA decapping complex, a key factor in the regulation of mRNA decay.Specifications
DCP1B | |
Polyclonal | |
Unconjugated | |
DCP1B | |
DCP1, hDcp1b, decapping enzyme hDcp1b, mRNA-decapping enzyme 1B | |
Rabbit | |
Antigen affinity chromatography | |
RUO | |
196513 | |
4° C | |
Liquid |
Immunoprecipitation | |
1 mg/ml | |
phosphate, tris citrate with 0.09% sodium azide; pH 7-8 | |
Q8IZD4 | |
DCP1B | |
Between 250 and 300 | |
100 μL | |
Primary | |
Human | |
Antibody | |
IgG |
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.