Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
DCP2 Antibody, DyLight 650, Novus Biologicals™

Rabbit Polyclonal Antibody
Supplier: Novus Biologicals NBP141070C
Description
DCP2 Polyclonal antibody specifically detects DCP2 in Human samples. It is validated for Western Blot, ImmunofluorescenceSpecifications
DCP2 | |
Polyclonal | |
Western Blot, Immunocytochemistry/Immunofluorescence | |
DCP2 decapping enzyme homolog (S. cerevisiae), EC 3.-, FLJ33245, hDpc, mRNA-decapping enzyme 2, nudix (nucleoside diphosphate linked moiety X)-type motif 20, Nudix motif 20, NUDT20Nucleoside diphosphate-linked moiety X motif 20 | |
The immunogen for this product maps to a region between residue 370 and 420 of human mRNA-decapping enzyme 2 using the numbering given in entry NP_689837.2 (GeneID 167227). | |
0.1 mL | |
Primary | |
Human | |
Purified |
Western Blot, Immunofluorescence | |
DyLight 650 | |
50mM Sodium Borate | |
Rabbit | |
Immunogen affinity purified | |
RUO | |
167227 | |
Store at 4°C in the dark. | |
IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction