Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
DCP2 Antibody (PCRP-DCP2-1D6), Biotin, Novus Biologicals™

Mouse Monoclonal Antibody
Supplier: Novus Biologicals NBP314037B
This item is not returnable.
View return policy
Description
DCP2 Monoclonal antibody specifically detects DCP2 in Human samples. It is validated for Flow Cytometry, ImmunofluorescenceSpecifications
| DCP2 | |
| Monoclonal | |
| Biotin | |
| DCP2 decapping enzyme homolog (S. cerevisiae), EC 3.-, FLJ33245, hDpc, mRNA-decapping enzyme 2, nudix (nucleoside diphosphate linked moiety X)-type motif 20, Nudix motif 20, NUDT20Nucleoside diphosphate-linked moiety X motif 20 | |
| Recombinant full-length human DCP2protein (Uniprot: Q8IU60) | |
| 0.1 mL | |
| Primary | |
| Human | |
| Purified |
| Flow Cytometry, Immunofluorescence | |
| PCRP-DCP2-1D6 | |
| PBS | |
| Mouse | |
| Protein A or G purified | |
| RUO | |
| 167227 | |
| Store at 4°C in the dark. | |
| IgG2a |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction