Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Dcp2 Rabbit anti-Mouse, Human, Polyclonal, MilliporeSigma™

Catalog No. ABE2901100 Shop All MilliporeSigma Products
Change view
Click to view available options
Quantity:
25 μL
100 μL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
ABE2901-100 100 μL
ABE2901-25UL 25 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. ABE2901-100 Supplier MilliporeSigma Supplier No. ABE2901100UL
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Anti-Dcp2 is a highly specific rabbit polyclonal antibody that targets m7GpppN-mRNA hydrolase / NUDT20 (Dcp2) and has been tested for use in Western Blotting.
TRUSTED_SUSTAINABILITY

Specifications

Antigen Dcp2
Applications Western Blot
Classification Polyclonal
Concentration Please refer to lot specific datasheet.
Conjugate Unconjugated
Gene Symbols DCP2
Host Species Rabbit
Immunogen His-tagged full length recombinant human m7GpppN-mRNA hydrolase (Dcp2).
Purification Method Unpurified
Quantity 100 μL
Regulatory Status RUO
Research Discipline Epigenetics and Nuclear Function
Primary or Secondary Primary
Gene ID (Entrez) NP_689837
Test Specificity This rabbit polyclonal antibody detects m7GpppN-mRNA hydrolase (Dcp2) in human and murine cells.
Target Species Mouse, Human
Content And Storage Stable for 1 year at -20°C from date of receipt. Handling Recommendations: Upon receipt and prior to removing the cap, centrifuge the vial and gently mix the solution. Aliquot into microcentrifuge tubes and store at -20°C. Avoid repeated freeze/thaw cycles, which may damage IgG and affect product performance.
Form Serum
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.