Learn More
Invitrogen™ DCPS Monoclonal Antibody (OTI1E6)

Description
DCPS Monoclonal Antibody for IHC (P), Western Blot
Specifications
Specifications
| Antigen | DCPS |
| Applications | Immunohistochemistry (Paraffin), Western Blot |
| Classification | Monoclonal |
| Clone | OTI1E6 |
| Concentration | 1 mg/mL |
| Conjugate | Unconjugated |
| Formulation | PBS with 1% BSA, 50% glycerol and 0.02% sodium azide; pH 7.3 |
| Gene | DCPS |
| Gene Accession No. | Q96C86 |
| Gene Alias | 5'-(N(7)-methyl 5'-triphosphoguanosine)-[mRNA] diphosphatase; ARS; Dcps; DCS1; DCS-1; decapping enzyme, scavenger; decapping scavenger enzyme; heat shock-like protein 1; HINT5; HINT-5; Hint-related 7meGMP-directed hydrolase; Histidine triad nucleotide-binding protein 5; histidine triad protein member 5; homolog of C. elegans 7meGMP-directed hydrolase dcs-1; HSL1; HSPC015; m7GpppX diphosphatase; mRNA decapping enzyme; scavenger mRNA-decapping enzyme DcpS |
| Show More |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.