Learn More
DCPS Mouse anti-Human, Clone: OTI1E6, lyophilized, TrueMAB™

Mouse Monoclonal Antibody
Supplier: OriGene CF505420

Description
Reconstitute with PBS (pH 7.3) and recommend to perform another round of desalting process using Product No. 7KMWCO
Necessary for the complete degradation of mRNAs, both in normal mRNA turnover and in nonsense-mediated mRNA decay. Removes the 7-methyl guanine cap structure from mRNA fragments shorter than 10 nucleotides that are produced by 3′->5′ exosome-mediated mRNA decay. Releases m7GMP. Can also degrade m7GDP to m7GMP. Has no activity towards mRNA molecules longer than 25 nucleotides.Specifications
DCPS | |
Monoclonal | |
Unconjugated | |
DCPS | |
ARS, DCS1, HINT-5, HINT5, HSL1, HSPC015 | |
Mouse | |
Affinity Chromatography | |
RUO | |
28960 | |
-20° C, Avoid Freeze/Thaw Cycles | |
Lyophilized |
Immunohistochemistry (Paraffin), Western Blot | |
OTI1E6 | |
PBS with 8% trehalose and no preservative; pH 7.3 | |
Q96C86 | |
DCPS | |
Full length human recombit protein of human DCPS produced in HEK293T cell | |
100 μg | |
Primary | |
Human | |
Antibody | |
IgG1 |
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.