Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ DCPS Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA530532
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA530532 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA530532 Supplier Invitrogen™ Supplier No. PA530532
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Recommended positive controls: A549. Predicted reactivity: Mouse (93%), Rat (94%), Pig (91%), Bovine (90%). Store product as a concentrated solution. Centrifuge briefly prior to opening the vial.

Necessary for the complete degradation of mRNAs, both in normal mRNA turnover and in nonsense-mediated mRNA decay. Removes the 7-methyl guanine cap structure from mRNA fragments shorter than 10 nucleotides that are produced by 3'->5' exosome-mediated mRNA decay. Releases m7GMP. Can also degrade m7GDP to m7GMP. Has no activity towards mRNA molecules longer than 25 nucleotides.
TRUSTED_SUSTAINABILITY

Specifications

Antigen DCPS
Applications Immunohistochemistry (Paraffin), Western Blot
Classification Polyclonal
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation 0.1M tris glycine with 20% glycerol and 0.01% thimerosal; pH 7
Gene DCPS
Gene Accession No. Q96C86
Gene Alias 5'-(N(7)-methyl 5'-triphosphoguanosine)-[mRNA] diphosphatase; ARS; Dcps; DCS1; DCS-1; decapping enzyme, scavenger; decapping scavenger enzyme; heat shock-like protein 1; HINT5; HINT-5; Hint-related 7meGMP-directed hydrolase; Histidine triad nucleotide-binding protein 5; histidine triad protein member 5; homolog of C. elegans 7meGMP-directed hydrolase dcs-1; HSL1; HSPC015; m7GpppX diphosphatase; mRNA decapping enzyme; scavenger mRNA-decapping enzyme DcpS
Gene Symbols DCPS
Host Species Rabbit
Immunogen Recombinant protein encompassing a sequence within the center region of human DCPS. The exact sequence is proprietary.
Purification Method Antigen affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 28960
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.