Learn More
Invitrogen™ DCPS Polyclonal Antibody

Description
Immunogen sequence: QLGKRKRELD VEEAHAASTE EKEAGVGNGT CAPVRLPFSG FRLQKVLRES ARDKIIFLHG KVNEASGDGD GEDAVVILEK TPFQVEQVAQ LLTGSPE Highest antigen sequence identity to the following orthologs: Mouse - 85%, Rat - 82%.
Specifications
Specifications
| Antigen | DCPS |
| Applications | Immunohistochemistry (Paraffin), Western Blot, Immunocytochemistry |
| Classification | Polyclonal |
| Concentration | 0.1 mg/mL |
| Conjugate | Unconjugated |
| Formulation | PBS with 40% glycerol and 0.02% sodium azide; pH 7.2 |
| Gene | DCPS |
| Gene Accession No. | Q96C86 |
| Gene Alias | 5'-(N(7)-methyl 5'-triphosphoguanosine)-[mRNA] diphosphatase; ARS; Dcps; DCS1; DCS-1; decapping enzyme, scavenger; decapping scavenger enzyme; heat shock-like protein 1; HINT5; HINT-5; Hint-related 7meGMP-directed hydrolase; Histidine triad nucleotide-binding protein 5; histidine triad protein member 5; homolog of C. elegans 7meGMP-directed hydrolase dcs-1; HSL1; HSPC015; m7GpppX diphosphatase; mRNA decapping enzyme; scavenger mRNA-decapping enzyme DcpS |
| Gene Symbols | DCPS |
| Show More |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.