Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ DcR3 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA523645
Change view
Click to view available options
Quantity:
400 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA523645 400 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA523645 Supplier Invitrogen™ Supplier No. PA523645
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

DcR3 Polyclonal Antibody for Western Blot, IHC (P), Flow

Apoptosis, or programmed cell death, occurs during normal cellular differentiation and development of multicellular organisms. Apoptosis is induced by certain cytokines including TNF (tumor necrosis factor) and Fas ligand of the TNF family through their death domain containing receptors, TNFR1 (tumor necrosis factor receptor 1) and Fas. Cell death signals are transduced by death domain (DD)-containing adapter molecules and members of the ICE (interleukin-1-beta converting enzyme)/CED-3 (C. elegans death receptor-3) protease family. A novel DD-containing molecule was recently cloned from mouse, human and monkey and designated Daxx (death associated protein 6). Daxx binds specifically to the Fas death domain and enhances Fas induced apoptosis and activates the Jun N-terminal kinase (JNK) pathway. Daxx is widely expressed in fetal and adult human and mouse tissues indicating its important function in Fas signaling pathways.
TRUSTED_SUSTAINABILITY

Specifications

Antigen DcR3
Applications Flow Cytometry, Immunohistochemistry (Paraffin), Western Blot
Classification Polyclonal
Concentration 0.5 mg/mL
Conjugate Unconjugated
Formulation PBS with 0.09% sodium azide; pH 7.4
Gene TNFRSF6B
Gene Accession No. O95407
Gene Alias DcR3; Decoy receptor 3; decoy receptor 3 variant 1; decoy receptor 3 variant 2; decoy receptor for Fas ligand; DJ583P15.1.1; hCG_2044836; M68; M68E; OTTHUMP00000031583; TNF receptor superfamily member 6b; TNFRSF6B; TR6; tumor necrosis factor receptor superfamily member 6B; tumor necrosis factor receptor superfamily member 6b, decoy; tumor necrosis factor receptor superfamily, member 6b, decoy; UNQ186/PRO212
Gene Symbols TNFRSF6B
Host Species Rabbit
Immunogen KLH conjugated synthetic peptide between 22-48 amino acids from the N-terminal region of human TNFRSF6B.
Purification Method Protein A, Antigen affinity chromatography
Quantity 400 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 8771
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.