Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Invitrogen™ DCST1 Monoclonal Antibody (2C12)
Mouse Monoclonal Antibody
Supplier: Invitrogen™ MA523086
Description
Peptide Sequence: MESNNMPCLP QPVGLDARAY WRAAVPIGLL VCLCLLQAFG YRLRRVIAAF YFPKREKKRI LFLYNDLLKK RAAFTKLRRA AILRRERQQK APRHPLADI.
This gene encodes a protein with a domain similar to one found in dendritic cells (PMID:11169400) which play a key role in antigen processing and display for immune responses. Multiple transcript variants encoding different isoforms have been found for this gene. [provided by RefSeq, Sep 2011].
Specifications
DCST1 | |
Monoclonal | |
1.0 mg/mL | |
PBS with no preservative; pH 7.4 | |
Q5T197 | |
DCST1 | |
DCST1 (NP_689707, 532 a.a. ~ 630 a.a) partial recombinant protein with GST tag. MW of the GST tag alone is 26 KDa. | |
100 μg | |
Primary | |
Human | |
Antibody | |
IgG2a κ |
ELISA | |
2C12 | |
Unconjugated | |
DCST1 | |
A330106H01Rik; Dcst1; DC-STAMP domain containing 1; DC-STAMP domain-containing protein 1; E3 ubiquitin-protein ligase DCST1; MLL5; RGD1307756; RING-type E3 ubiquitin transferase | |
Mouse | |
Affinity chromatography | |
RUO | |
149095 | |
-20°C, Avoid Freeze/Thaw Cycles | |
Liquid |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction