Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

DCST1 Rabbit anti-Human, Polyclonal, Bioss
SDP

Catalog No. 501988402
Encompass_Preferred
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
50-198-8402 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 50-198-8402 Supplier Bioss Supplier No. BS8250R
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

This gene encodes a protein with a domain similar to one found in dendritic cells (PMID:11169400) which play a key role in antigen processing and display for immune responses. Multiple transcript variants encoding different isoforms have been found for this gene. [provided by RefSeq, Sep 2011].

Specifications

Antigen DCST1
Applications Immunofluorescence, Immunohistochemistry (Paraffin), Western Blot
Classification Polyclonal
Concentration 1 μg/mL
Conjugate Unconjugated
Formulation PBS with 1% BSA, 50% glycerol and 0.09% sodium azide; pH
Gene DCST1
Gene Accession No. Q5T197
Gene Alias BOS_2886; DCST1; DC-STAMP domain containing 1; DC-STAMP domain-containing protein 1; E3 ubiquitin-protein ligase DCST1; MLL5; RGD1307756; RING-type E3 ubiquitin transferase
Gene Symbols DCST1
Host Species Rabbit
Immunogen KLH conjugated synthetic peptide derived from human DCST1.
Purification Method Protein A
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 149095
Target Species Human
Content And Storage -20°C
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.