Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

DCT Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA5101647
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA5101647 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA5101647 Supplier Thermo Scientific Supplier No. PA5101647
Add to Cart
Add to Cart

DCT Polyclonal Antibody

DCT Polyclonal Antibody for Western Blot, ICC/IF

TRUSTED_SUSTAINABILITY

Specifications

Antigen DCT
Applications Western Blot, Immunocytochemistry
Classification Polyclonal
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation PBS with 50% glycerol and 0.02% sodium azide; pH 7.4
Gene DCT
Gene Accession No. P29812, P40126
Gene Alias Dct; DOPAchrome conversion factor; dopachrome delta isomerase; dopachrome delta-isomerase; DOPAchrome isomerase; DOPAchrome oxidoreductase; dopachrome tautomerase; dopachrome tautomerase (dopachrome delta-isomerase, tyrosine-related protein 2); DT; hypothetical protein LOC664745; L-dopachrome Delta-isomerase; L-dopachrome isomerase; L-dopachrome tautomerase; melanogenic enzyme; tyrosinase-related protein 2; trp2; RGD1564975; slaty; SLATY locus protein; slt; trp2; TRP-2; tyrosinase related protein 2; tyrosinase related protein-2; tyrosinase-related protein 2; tyrosinase-related protein-2; tyrosine-related protein 2; Tyrp2; Tyrp-2; zgc:109860
Gene Symbols DCT
Host Species Rabbit
Immunogen A synthesized peptide derived from human DCT(Accession P40126), corresponding to amino acid residues Q56-C106.
Purification Method Affinity Chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 13190, 1638
Target Species Human, Mouse
Content And Storage -20°C
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.