Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

DCT Polyclonal Antibody, Invitrogen™

Catalog No. PIPA536485
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA536485 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA536485 Supplier Thermo Scientific Supplier No. PA536485

Item Discontinued This item has been discontinued by the supplier. Please to view product availability in your area.

Rabbit Polyclonal Antibody

This antibody detects endogenous protein at a molecular weight of 60 kDa. Purity is >95% by SDS-PAGE.

DCT Polyclonal antibody specifically detects DCT in Human, Mouse, Rat samples. It is validated for Immunocytochemistry, Immunohistochemistry.
TRUSTED_SUSTAINABILITY

Specifications

Antigen DCT
Applications Immunocytochemistry, Immunohistochemistry
Classification Polyclonal
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation PBS with 50% glycerol and 0.02% sodium azide; pH 7.2
Gene DCT
Gene Accession No. P29812, P40126
Gene Alias Dct; DOPAchrome conversion factor; dopachrome delta isomerase; dopachrome delta-isomerase; DOPAchrome isomerase; DOPAchrome oxidoreductase; dopachrome tautomerase; dopachrome tautomerase (dopachrome delta-isomerase, tyrosine-related protein 2); DT; hypothetical protein LOC664745; L-dopachrome Delta-isomerase; L-dopachrome isomerase; L-dopachrome tautomerase; melanogenic enzyme; tyrosinase-related protein 2; trp2; RGD1564975; slaty; SLATY locus protein; slt; trp2; TRP-2; tyrosinase related protein 2; tyrosinase related protein-2; tyrosinase-related protein 2; tyrosinase-related protein-2; tyrosine-related protein 2; Tyrp2; Tyrp-2; zgc:109860
Gene Symbols DCT
Host Species Rabbit
Immunogen Synthetic peptide corresponding to amino acids 50-100 of human TRP2
Purification Method Antigen affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 13190, 1638, 290484
Target Species Human, Mouse, Rat
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.