Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

DCTN5 Rabbit anti-Human, Polyclonal, Bioss
SDP

Catalog No. 501983245
Encompass_Preferred
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
50-198-3245 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 50-198-3245 Supplier Bioss Supplier No. BS14308R
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

This gene encodes a subunit of dynactin, a component of the cytoplasmic dynein motor machinery involved in minus-end-directed transport. The encoded protein is a component of the pointed-end subcomplex and is thought to bind membranous cargo. A pseudogene of this gene is located on the long arm of chromosome 1. Alternatively spliced transcript variants encoding multiple isoforms have been observed for this gene.

Specifications

Antigen DCTN5
Applications Immunohistochemistry (Paraffin)
Classification Polyclonal
Concentration 1 μg/mL
Conjugate Unconjugated
Formulation PBS with 1% BSA, 50% glycerol and 0.09% sodium azide; pH
Gene DCTN5
Gene Accession No. Q9BTE1
Gene Alias 4930427E12Rik; b2b315Clo; BOS_22751; C78178; Dctn5; dynactin 4; dynactin 5; dynactin 5 (p25); dynactin 5-like; Dynactin subunit 5; Dynactin subunit p25; LOC100349573; p25; p25 dynactin subunit; RGD1305654; zgc:92842
Gene Symbols DCTN5
Host Species Rabbit
Immunogen KLH conjugated synthetic peptide derived from human DCTN5.
Purification Method Protein A
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 84516
Target Species Human
Content And Storage -20°C
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.