Learn More
Invitrogen™ DCTN6 Polyclonal Antibody

Description
The antibody detects endogenous levels of total DCTN6 protein.
Specifications
Specifications
| Antigen | DCTN6 |
| Applications | Immunohistochemistry (Paraffin), Western Blot |
| Classification | Polyclonal |
| Concentration | 2.6 mg/mL |
| Conjugate | Unconjugated |
| Formulation | PBS with 40% glycerol and 0.05% sodium azide; pH 7.4 |
| Gene | DCTN6 |
| Gene Accession No. | O00399 |
| Gene Alias | AU044699; Dctn6; dynactin 6; dynactin subunit 6; Dynactin subunit p27; novel RGD-containing protein; p27; p27 dynactin subunit; protein WS-3; RGD containing protein; WS3; WS-3 |
| Gene Symbols | DCTN6 |
| Show More |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.