Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ DCTN6 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA597003
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA597003 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA597003 Supplier Invitrogen™ Supplier No. PA597003
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Immunogen sequence: MAEKTQKSVK IAPGAVVCVE SEIRGDVTIG PRTVIHPKAR IIAEAGPIVI GEGNLIEEQA LIINAYPDNI TPDTEDPEPK PMIIGTNNVF EVGCYSQAMK MGDNNVIESK AYVGRNVILT SGCIIGACCN LNTFEVIPEN TVIYGADCLR RVQTERPQPQ TLQLDFLMKI LPNYHHLKKT MKGSSTPVKN; Positive Samples: A-549, U-251MG, 22Rv1, Mouse brain, Mouse thymus; Cellular Location: Chromosome, Cytoplasm, centromere, cytoskeleton, kinetochore.

The protein encoded by this gene contains an RGD (Arg-Gly-Asp) motif in the N-terminal region, which confers adhesive properties to macromolecular proteins like fibronectin. It shares a high degree of sequence similarity with the mouse homolog, which has been suggested to play a role in mitochondrial biogenesis. The exact biological function of this gene is not known. [provided by RefSeq, Jul 2008].
TRUSTED_SUSTAINABILITY

Specifications

Antigen DCTN6
Applications ELISA, Western Blot
Classification Polyclonal
Concentration 3.03 mg/mL
Conjugate Unconjugated
Formulation PBS with 50% glycerol and 0.02% sodium azide; pH 7.3
Gene DCTN6
Gene Accession No. O00399, Q9WUB4
Gene Alias AU044699; Dctn6; dynactin 6; dynactin subunit 6; Dynactin subunit p27; novel RGD-containing protein; p27; p27 dynactin subunit; protein WS-3; RGD containing protein; WS3; WS-3
Gene Symbols DCTN6
Host Species Rabbit
Immunogen Recombinant fusion protein containing a sequence corresponding to amino acids 1-190 of human DCTN6 (NP_0065621).
Purification Method Affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 10671, 22428
Target Species Human, Mouse
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.