Learn More
Invitrogen™ DcTRAILR1 Monoclonal Antibody (270012)
Rat Monoclonal Antibody
Supplier: Invitrogen™ MA523971
Description
In Western blots, less than 10% cross-reactivity with recombinant mouse (rm) 4-1BB Ligand, recombinant human (rh) BAFF R, rhDcTRAIL R3, rmFas, rmOPG, and rhTRAIL R4 is observed and no cross-reactivity with rmCD27, rmCD30, rmDR3, rhDR6, rmEDAR, recombinant feline Fas, rhFas, recombinant rat Fas, rmGITR, rmHVEM, rmLymphotoxin beta R, rmNGF R, rmOX40, rmTNF RI, rmTNF RII, rhTRAIL R1, or rhTROY is observed. Reconstitute at 0.5 mg/mL in sterile PBS.
Mouse DcTRAILR1 and DcTRAILR2 (decoy TRAIL receptors 1 and 2) proteins are receptors for TRAIL according to ligand-receptor binding assays (Schneider et al. 2001). However, overexpression does not cause NF-kB activation or apoptosis, as would be expected of a TRAIL receptor. These observations led researchers to refer to these receptors as decoys. It is thought that cells that express decoy TRAIL receptor decoys at high levels relative to actual TRAIL receptors may use decoys as protection against the cytotoxic action of TRAIL (reviewed in Bouralexis et al. 2005). Additional analysis of these two murine TRAIL decoy receptors should help faciliate the understanding of the complex biology underlying TRAIL ligand/receptor interactions. DcTRAILR1 is an amino acid protein of 176 amino acids (GenBank no: NP-077252.2). DcTRAILR2 is an amino acid protein of 198 amino acids (GenBank no: AAN87807.1).
Specifications
DcTRAILR1 | |
Monoclonal | |
0.5 mg/mL | |
PBS with 5% trehalose and No Preservative | |
Q9ER63 | |
Tnfrsf23 | |
Mouse myeloma cell line NS0-derived recombinant mouse DcTRAIL R1/TNFRSF23 Met30-Ser156. | |
100 μg | |
Primary | |
Mouse | |
Antibody | |
IgG2a |
Western Blot, Immunocytochemistry | |
270012 | |
Unconjugated | |
Tnfrsf23 | |
Dctrailr1; Decoy TRAIL receptor 1; mDcTRAILR1; MGC123882; MGC123883; mSOB; SOB; TNF receptor family member SOB; TNF receptor homolog 1; Tnfrh1; Tnfrsf1al1; Tnfrsf23; TNFRSFH23; Tumor necrosis factor receptor p60 homolog 1; Tumor necrosis factor receptor superfamily member 23; tumor necrosis factor receptor superfamily, member 23 | |
Rat | |
Protein A/G | |
RUO | |
79201 | |
-20°C, Avoid Freeze/Thaw Cycles | |
Lyophilized |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.