Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
DCUN1D1 Antibody, Alexa Fluor™ 647, Novus Biologicals™

Rabbit Polyclonal Antibody
Supplier: Novus Biologicals NBP296995AF647
Description
DCUN1D1 Polyclonal specifically detects DCUN1D1 in Human samples. It is validated for Western Blot, ELISA.Specifications
DCUN1D1 | |
Polyclonal | |
Western Blot, ELISA | |
DCN1, defective in cullin neddylation 1, domain containing 1 (S. cerevisiae), DCN1-like protein 1, DCUN1L1DCUN1 domain-containing protein 1, Defective in cullin neddylation protein 1-like protein 1, RP42 homolog, RP42Squamous cell carcinoma-related oncogene, SCCROSCRO, Tes3 | |
Produced in rabbits immunized with purified, recombinant DCUN1D1 (NP_065691.2; Met1-Val259). | |
0.1 mL | |
Cancer | |
54165 | |
Store at 4°C in the dark. | |
IgG |
Western Blot, ELISA | |
Alexa Fluor 647 | |
50mM Sodium Borate | |
Rabbit | |
Affinity purified | |
RUO | |
Primary | |
Human | |
Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction