Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
DCUN1D1 Antibody, Novus Biologicals™

Rabbit Polyclonal Antibody
Supplier: Novus Biologicals NBP216116
Description
DCUN1D1 Polyclonal specifically detects DCUN1D1 in Human samples. It is validated for Western Blot.Specifications
DCUN1D1 | |
Polyclonal | |
Unconjugated | |
0.1M Tris (pH 7.0), 0.1M Glycine and 20% Glycerol with 0.01% Thimerosal | |
DCN1, defective in cullin neddylation 1, domain containing 1 (S. cerevisiae), DCN1-like protein 1, DCUN1L1DCUN1 domain-containing protein 1, Defective in cullin neddylation protein 1-like protein 1, RP42 homolog, RP42Squamous cell carcinoma-related oncogene, SCCROSCRO, Tes3 | |
Rabbit | |
30 kDa | |
0.1 mL | |
Cancer | |
54165 | |
Human | |
IgG |
Western Blot | |
0.24mg/mL | |
Western Blot 1:500-1:3000 | |
Q96GG9 | |
DCUN1D1 | |
Recombinant protein encompassing a sequence within the center region of human DCUN1D1. The exact sequence is proprietary. | |
Affinity Purified | |
RUO | |
Primary | |
Human. Expected to cross react based on sequence identity: Mouse (99%), Chicken (98%), Xenopus laevis (97%), Zebrafish (88%). | |
Aliquot and store at -20C or -80C. Avoid freeze-thaw cycles. |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction