Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Invitrogen™ DCUN1D1 Polyclonal Antibody
Rabbit Polyclonal Antibody
Supplier: Invitrogen™ PA580543
Description
This product is preservative free. It is recommended to add sodium azide to avoid contamination (final concentration 0.05%-0.1%). This antibody has specificity for Human DCUN1D1/SCCRO.
Part of an E3 ubiquitin ligase complex for neddylation. Promotes neddylation of cullin components of E3 cullin-RING ubiquitin ligase complexes. Acts by binding to cullin-RBX1 complexes in the cytoplasm and promoting their nuclear translocation, enhancing recruitment of E2-NEDD8 (UBE2M-NEDD8) thioester to the complex, and optimizing the orientation of proteins in the complex to allow efficient transfer of NEDD8 from the E2 to the cullin substrates. Involved in the release of inhibitory effets of CAND1 on cullin-RING ligase E3 complex assembly and activity (PubMed:25349211, PubMed:28581483). Acts also as an oncogene facilitating malignant transformation and carcinogenic progression. [UniProt].
Specifications
DCUN1D1 | |
Polyclonal | |
Unconjugated | |
DCUN1D1 | |
DCN1; DCN1, defective in cullin neddylation 1, domain containing 1; DCN1, defective in cullin neddylation 1, domain containing 1 (S. cerevisiae); DCN1-like protein 1; DCNL1; DCUN1 domain-containing protein 1; Dcun1d1; DCUN1L1; defective in cullin neddylation 1 domain containing 1; defective in cullin neddylation protein 1-like protein 1; pTes3; Rp42; RP42 homolog; SCCRO; SCRO; squamous cell carcinoma related oncogene; squamous cell carcinoma-related oncogene; Tes3; testis derived transcript 3; testis-specific protein 3 | |
Rabbit | |
Antigen affinity chromatography, Protein A | |
RUO | |
54165 | |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
Liquid |
ELISA, Immunoprecipitation, Western Blot | |
1 mg/mL | |
PBS with no preservative | |
Q96GG9 | |
DCUN1D1 | |
Recombinant Human DCUN1D1/SCCRO protein (Met1-Val259). | |
100 μL | |
Primary | |
Human | |
Antibody | |
IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction