Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ DCUN1D1 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA583298
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA583298 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA583298 Supplier Invitrogen™ Supplier No. PA583298
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Immunogen sequence: RQFMIFTQSS EKTAVSCLSQ NDWKLDVATD NFFQNPELYI RESVKGSLDR KKLEQLYNR Highest antigen sequence indentity to the following orthologs: Rat - 100%, Mouse - 98%.

Part of an E3 ubiquitin ligase complex for neddylation. Promotes neddylation of cullin components of E3 cullin-RING ubiquitin ligase complexes. Acts by binding to cullin-RBX1 complexes in the cytoplasm and promoting their nuclear translocation, enhancing recruitment of E2-NEDD8 (UBE2M-NEDD8) thioester to the complex, and optimizing the orientation of proteins in the complex to allow efficient transfer of NEDD8 from the E2 to the cullin substrates. Involved in the release of inhibitory effets of CAND1 on cullin-RING ligase E3 complex assembly and activity (PubMed:25349211, PubMed:28581483). Acts also as an oncogene facilitating malignant transformation and carcinogenic progression. [UniProt].
TRUSTED_SUSTAINABILITY

Specifications

Antigen DCUN1D1
Applications Immunohistochemistry (Paraffin), Western Blot, Immunocytochemistry
Classification Polyclonal
Concentration 0.05 mg/mL
Conjugate Unconjugated
Formulation PBS with 40% glycerol and 0.02% sodium azide; pH 7.2
Gene DCUN1D1
Gene Accession No. Q96GG9
Gene Alias DCN1; DCN1, defective in cullin neddylation 1, domain containing 1; DCN1, defective in cullin neddylation 1, domain containing 1 (S. cerevisiae); DCN1-like protein 1; DCNL1; DCUN1 domain-containing protein 1; Dcun1d1; DCUN1L1; defective in cullin neddylation 1 domain containing 1; defective in cullin neddylation protein 1-like protein 1; pTes3; Rp42; RP42 homolog; SCCRO; SCRO; squamous cell carcinoma related oncogene; squamous cell carcinoma-related oncogene; Tes3; testis derived transcript 3; testis-specific protein 3
Gene Symbols DCUN1D1
Host Species Rabbit
Immunogen Recombinant protein corresponding to Human DCUN1D1. Recombinant protein control fragment (Product #RP-96262).
Purification Method Antigen affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 54165
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.