Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
DCUN1D2 Antibody, mFluor Violet 450 SE, Novus Biologicals™

Rabbit Polyclonal Antibody
Supplier: Novus Biologicals NBP297034MFV450
Description
DCUN1D2 Polyclonal antibody specifically detects DCUN1D2 in Human samples. It is validated for Western Blot, ELISA, ImmunoprecipitationSpecifications
DCUN1D2 | |
Polyclonal | |
Western Blot, ELISA, Immunoprecipitation | |
C13orf17, chromosome 13 open reading frame 17, DCN1, defective in cullin neddylation 1, domain containing 2 (S. cerevisiae), DCN1-like protein 2, DCUN1 domain-containing protein 2, DCUN1L2, Defective in cullin neddylation protein 1-like protein 2, FLJ10704, FLJ20092 | |
Produced in rabbits immunized with purified, recombinant Human DCUN1D2 (Accession#: NP_001014305.1; Met1-Phe259) | |
0.1 mL | |
Primary | |
Human | |
Purified |
Western Blot, ELISA, Immunoprecipitation | |
mFluor Violet 450 SE | |
50mM Sodium Borate | |
Rabbit | |
Antigen and protein A Affinity-purified | |
RUO | |
55208 | |
Store at 4C in the dark. | |
IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction