Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Invitrogen™ DCUN1D2 Polyclonal Antibody
Rabbit Polyclonal Antibody
Supplier: Invitrogen™ PA531607
Description
Recommended positive controls: A549, HeLa, HepG2, HCT116. Predicted reactivity: Mouse (94%), Rat (94%), Xenopus laevis (89%). Store product as a concentrated solution. Centrifuge briefly prior to opening the vial.
Potently stimulates the neddylation of cullin components of SCF-type E3 ubiquitin ligase complexes from the NEDD8-conjugating E2 enzyme UBC12. Neddylation of cullins play an essential role in the regulation of SCF-type complexes activity. [UniProt].
Specifications
DCUN1D2 | |
Polyclonal | |
Unconjugated | |
DCUN1D2 | |
AW495713; C13orf17; DCN1, defective in cullin neddylation 1, domain containing 2; DCN1, defective in cullin neddylation 1, domain containing 2 (S. cerevisiae); DCN1-like protein 2; DCN1-like protein 2; LOW QUALITY PROTEIN: DCN1-like protein 2; DCUN1 domain-containing protein 2; Dcun1d2; DCUN1L2; defective in cullin neddylation 1 domain containing 2; defective in cullin neddylation protein 1-like protein 2; mFLJ10704; RGD1311743; squamous cell carcinoma-related oncogene | |
Rabbit | |
Antigen Affinity Chromatography | |
RUO | |
55208 | |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
Liquid |
Immunohistochemistry (Paraffin), Western Blot | |
0.97 mg/mL | |
0.1M tris glycine with 20% glycerol and 0.01% thimerosal; pH 7 | |
Q6PH85 | |
DCUN1D2 | |
Recombinant fragment corresponding to a region within amino acids 52 and 259 of Human DCUN1D2. | |
100 μL | |
Primary | |
Human | |
Antibody | |
IgG |
Safety and Handling
WARNING: Cancer - www.P65Warnings.ca.gov
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction