Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Invitrogen™ DCUN1D2 Polyclonal Antibody
Rabbit Polyclonal Antibody
Supplier: Invitrogen™ PA558748
Description
Immunogen sequence: QFMACTQAGE RTAIYCLTQN EWRLDEATDS FFQNPDSLHR ESMRNAVDKK KLERLYGRY Highest antigen sequence identity to the following orthologs: Mouse - 81%, Rat - 80%.
Potently stimulates the neddylation of cullin components of SCF-type E3 ubiquitin ligase complexes from the NEDD8-conjugating E2 enzyme UBC12. Neddylation of cullins play an essential role in the regulation of SCF-type complexes activity. [UniProt].
Specifications
DCUN1D2 | |
Polyclonal | |
Unconjugated | |
DCUN1D2 | |
AW495713; C13orf17; DCN1, defective in cullin neddylation 1, domain containing 2; DCN1, defective in cullin neddylation 1, domain containing 2 (S. cerevisiae); DCN1-like protein 2; DCN1-like protein 2; LOW QUALITY PROTEIN: DCN1-like protein 2; DCUN1 domain-containing protein 2; Dcun1d2; DCUN1L2; defective in cullin neddylation 1 domain containing 2; defective in cullin neddylation protein 1-like protein 2; mFLJ10704; RGD1311743; squamous cell carcinoma-related oncogene | |
Rabbit | |
Antigen affinity chromatography | |
RUO | |
55208 | |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
Liquid |
Immunohistochemistry (Paraffin), Immunocytochemistry | |
0.4 mg/mL | |
PBS with 40% glycerol and 0.02% sodium azide; pH 7.2 | |
Q6PH85 | |
DCUN1D2 | |
Recombinant protein corresponding to Human DCUN1D2. Recombinant protein control fragment (Product #RP-97975). | |
100 μL | |
Primary | |
Human | |
Antibody | |
IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction