Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

DCUN1D2 Rabbit anti-Human, Polyclonal, Bioss
SDP

Catalog No. 501986917
Encompass_Preferred
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
50-198-6917 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 50-198-6917 Supplier Bioss Supplier No. BS5931R
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

May contribute to neddylation of cullin components of SCF-type E3 ubiquitin ligase complexes. Neddylation of cullins play an essential role in the regulation of SCF-type complexes activity.

Specifications

Antigen DCUN1D2
Applications ELISA, Immunofluorescence, Immunohistochemistry (Paraffin), Western Blot
Classification Polyclonal
Concentration 1 μg/mL
Conjugate Unconjugated
Formulation PBS with 1% BSA, 50% glycerol and 0.09% sodium azide; pH
Gene DCUN1D2
Gene Accession No. Q6PH85
Gene Alias AW495713; C13orf17; DCN1, defective in cullin neddylation 1, domain containing 2; DCN1, defective in cullin neddylation 1, domain containing 2 (S. cerevisiae); DCN1, defective in cullin neddylation 1, domain containing 2-like; DCN1-like protein 2; DCUN1 domain-containing protein 2; DCUN1D2; Dcun1l2; defective in cullin neddylation 1 domain containing 2; Defective in cullin neddylation protein 1-like protein 2; LOC100355008; mFLJ10704; RGD1311743; squamous cell carcinoma-related oncogene
Gene Symbols DCUN1D2
Host Species Rabbit
Immunogen KLH conjugated synthetic peptide derived from human DCUN1D1.
Purification Method Protein A
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 55208
Target Species Human
Content And Storage -20°C
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.