Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ DCUN1D4 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA544179
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA544179 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA544179 Supplier Invitrogen™ Supplier No. PA544179
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Peptide sequence: TLNKLNLTED IGQDDHQTGS LRSCSSSDCF NKVMPPRKKR RPASGDDLSA Sequence homology: Cow: 93%; Dog: 86%; Guinea Pig: 93%; Horse: 93%; Human: 100%; Mouse: 93%; Rabbit: 86%; Rat: 93%.

DCUN1D4 contains 1 DCUN1 domain. The exact function of DCUN1D4 remains unknown.
TRUSTED_SUSTAINABILITY

Specifications

Antigen DCUN1D4
Applications Western Blot
Classification Polyclonal
Concentration 0.5 mg/mL
Conjugate Unconjugated
Formulation PBS with 2% sucrose and 0.09% sodium azide
Gene DCUN1D4
Gene Accession No. Q92564
Gene Alias AI836376; DCN1, defective in cullin neddylation 1, domain containing 4; DCN1, defective in cullin neddylation 1, domain containing 4 (S. cerevisiae); DCN1-like protein 4; DCUN1 domain-containing protein 4; Dcun1d4; defective in cullin neddylation 1 domain containing 4; defective in cullin neddylation protein 1-like protein 4; KIAA0276; RGD1310422; Unknown (protein for MGC:134196)
Gene Symbols DCUN1D4
Host Species Rabbit
Immunogen Synthetic peptide directed towards the N-terminal of human DCUN1D4 (aa 28-77).
Purification Method Affinity Chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 23142
Target Species Human
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.