Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Invitrogen™ DCUN1D4 Polyclonal Antibody

Rabbit Polyclonal Antibody
Supplier: Invitrogen™ PA557681
Description
Immunogen sequence: FNKVMPPRKK RRPASGDDLS AKKSRHDSMY RKYDSTRIKT EEEAFSSKRC LEWFYE Highest antigen sequence identity to the following orthologs: Mouse - 96%, Rat - 96%.
DCUN1D4 contains 1 DCUN1 domain. The exact function of DCUN1D4 remains unknown.
Specifications
DCUN1D4 | |
Polyclonal | |
Unconjugated | |
DCUN1D4 | |
AI836376; DCN1, defective in cullin neddylation 1, domain containing 4; DCN1, defective in cullin neddylation 1, domain containing 4 (S. cerevisiae); DCN1-like protein 4; DCUN1 domain-containing protein 4; Dcun1d4; defective in cullin neddylation 1 domain containing 4; defective in cullin neddylation protein 1-like protein 4; KIAA0276; RGD1310422; Unknown (protein for MGC:134196) | |
Rabbit | |
Antigen affinity chromatography | |
RUO | |
100737, 23142, 360928 | |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
Liquid |
Immunohistochemistry (Paraffin), Western Blot, Immunocytochemistry | |
0.1 mg/mL | |
PBS with 40% glycerol and 0.02% sodium azide; pH 7.2 | |
Q8CCA0, Q92564 | |
DCUN1D4 | |
Recombinant protein corresponding to Human DCUN1D4. Recombinant protein control fragment (Product #RP-96617). | |
100 μL | |
Primary | |
Human, Mouse, Rat | |
Antibody | |
IgG |
Safety and Handling
WARNING: Cancer - www.P65Warnings.ca.gov
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction