Learn More
DCX Mouse anti-Human, Clone: OTI4A3, lyophilized, TrueMAB™

Mouse Monoclonal Antibody
Supplier: OriGene CF501778

Description
Reconstitute with PBS (pH 7.3) and recommend to perform another round of desalting process using Product No. 7KMWCO
In the developing cortex, cortical neurons must migrate over long distances to reach the site of their final differentiation. DCX is a cytoplasmic protein which appears to direct neuronal migration by regulating the organization and stability of microtubules. It contains two doublecortin domains, which bind microtubules. In addition, this protein interacts with LIS1, the regulatory gamma subunit of platelet activating factor acetylhydrolase, and this interaction is important to proper microtubule function in the developing cortex. Mutations in the gene encoding DCX are a cause of X-linked lissencephaly.Specifications
DCX | |
Monoclonal | |
Unconjugated | |
DCX | |
DBCN, DC, LISX, SCLH, XLIS | |
Mouse | |
Affinity Chromatography | |
RUO | |
1641 | |
-20° C, Avoid Freeze/Thaw Cycles | |
Lyophilized |
Flow Cytometry, Immunocytochemistry, Immunofluorescence, Western Blot | |
OTI4A3 | |
PBS with 8% trehalose and no preservative; pH 7.3 | |
O43602 | |
DCX | |
Full length human recombit protein of human DCX produced in HEK293T cell. | |
100 μg | |
Primary | |
Human | |
Antibody | |
IgG1 |
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.