Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
DCXR Antibody (OTI7D11), DyLight 550, Novus Biologicals™

Mouse Monoclonal Antibody
Supplier: Novus Biologicals NBP271906R
Description
DCXR Monoclonal specifically detects DCXR in Human samples. It is validated for Western Blot, Immunohistochemistry.Specifications
DCXR | |
Monoclonal | |
DyLight 550 | |
50mM Sodium Borate with 0.05% Sodium Azide | |
DCXR | |
Full length recombinant protein of human DCXR (NP_057370) produced in HEK293T cells. | |
0.1 mL | |
Primary | |
Human | |
IgG1 |
Western Blot, Immunohistochemistry | |
OTI7D11 | |
Western Blot, Immunohistochemistry | |
Carbonyl reductase II, DCR, dicarbonyl/L-xylulose reductaseP34H, EC 1.1.1, EC 1.1.1.10, HCR2, HCRII, human carbonyl reductase 2, KIDCR, Kidney dicarbonyl reductase, L-xylulose reductase, SDR20C1, short chain dehydrogenase/reductase family 20C, member 1, Sperm surface protein P34H, XR | |
Mouse | |
Affinity Purified | |
Apoptosis, Cell Biology, Cytokine Research, Immunology, Innate Immunity, Lipid and Metabolism | |
51181.0 | |
Store at 4°C in the dark. |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction