Learn More
Invitrogen™ DCXR Monoclonal Antibody (OTI4D11)

Description
DCXR Monoclonal Antibody for Western Blot, ICC/IF, IHC (P), Flow
Specifications
Specifications
| Antigen | DCXR |
| Applications | Flow Cytometry, Immunohistochemistry (Paraffin), Western Blot, Immunocytochemistry |
| Classification | Monoclonal |
| Clone | OTI4D11 |
| Concentration | 0.53 mg/mL |
| Conjugate | Unconjugated |
| Formulation | PBS with 1% BSA, 50% glycerol and 0.02% sodium azide; pH 7.3 |
| Gene | DCXR |
| Gene Accession No. | Q7Z4W1 |
| Gene Alias | 0610038K04Rik; 1810027P18Rik; carbonyl reductase; carbonyl reductase 2; carbonyl reductase II; DCR; Dcxr; diacetyl/L-xylulose reductase; dicarbonyl and L-xylulose reductase; dicarbonyl L-xylulose reductase; dicarbonyl/L-xylulose reductase; Glb; HCR2; HCRII; KI P34H; KID; KIDCR; Kidney dicarbonyl reductase; L-xylulose reductase; P34H; PNTSU; SDR20C1; Short chain dehydrogenase/reductase family 20C member 1; short chain dehydrogenase/reductase family 20C, member 1; sperm surface protein P34H; XR |
| Show More |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.