Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
DCXR Mouse anti-Human, Clone: OTI7D11, liquid, TrueMAB™

Mouse Monoclonal Antibody
Supplier: OriGene TA500773

Description
The protein encoded by this gene acts as a homotetramer to catalyze diacetyl reductase and L-xylulose reductase reactions. The encoded protein may play a role in the uronate cycle of glucose metabolism and in the cellular osmoregulation in the proximal renal tubules. Defects in this gene are a cause of pentosuria. Two transcript variants encoding different isoforms have been found for this gene.Specifications
DCXR | |
Monoclonal | |
0.51 mg/mL | |
PBS with 1% BSA, 50% glycerol and 0.02% sodium azide | |
Q7Z4W1 | |
DCXR | |
Full length human recombit protein of human DCXR produced in HEK293T cell. | |
100 μL | |
Primary | |
Human | |
Antibody | |
IgG1 |
Immunohistochemistry (Paraffin), Western Blot | |
OTI7D11 | |
Unconjugated | |
DCXR | |
DCR, HCR2, HCRII, KIDCR, P34H, PNTSU, SDR20C1, XR | |
Mouse | |
Affinity Chromatography | |
RUO | |
51181 | |
-20° C, Avoid Freeze/Thaw Cycles | |
Liquid |
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction