Learn More
DCXR Antibody (OTI7D11), DyLight 350, Novus Biologicals™
SDP
Mouse Monoclonal Antibody
Supplier:  Novus Biologicals NBP271906UV
Catalog No. NBP271906UV
Only null left
Add to Cart
Description
Description
DCXR Monoclonal specifically detects DCXR in Human samples. It is validated for Western Blot, Immunohistochemistry.
Specifications
Specifications
DCXR
Monoclonal
DyLight 350
50mM Sodium Borate with 0.05% Sodium Azide
DCXR
Full length recombinant protein of human DCXR (NP_057370) produced in HEK293T cells.
0.1 mL
Primary
Human
IgG1
Western Blot, Immunohistochemistry
OTI7D11
Western Blot, Immunohistochemistry
Carbonyl reductase II, DCR, dicarbonyl/L-xylulose reductaseP34H, EC 1.1.1, EC 1.1.1.10, HCR2, HCRII, human carbonyl reductase 2, KIDCR, Kidney dicarbonyl reductase, L-xylulose reductase, SDR20C1, short chain dehydrogenase/reductase family 20C, member 1, Sperm surface protein P34H, XR
Mouse
Affinity Purified
Apoptosis, Cell Biology, Cytokine Research, Immunology, Innate Immunity, Lipid and Metabolism
51181.0
Store at 4°C in the dark.
Product Suggestions
Product Suggestions
Videos
SDS
Documents
Documents
Product Certifications
Promotions
Promotions
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.