Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ DCXR Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA518071
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA518071 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA518071 Supplier Invitrogen™ Supplier No. PA518071
Add to Cart
Add to Cart

Goat Polyclonal Antibody

This antibody is tested in Peptide ELISA: antibody detection limit dilution 32,000.

The protein encoded by this gene acts as a homotetramer to catalyze diacetyl reductase and L-xylulose reductase reactions. The encoded protein may play a role in the uronate cycle of glucose metabolism and in the cellular osmoregulation in the proximal renal tubules. Defects in this gene are a cause of pentosuria. Two transcript variants encoding different isoforms have been found for this gene.
TRUSTED_SUSTAINABILITY

Specifications

Antigen DCXR
Applications Immunohistochemistry (Paraffin), Western Blot
Classification Polyclonal
Concentration 0.5 mg/mL
Conjugate Unconjugated
Formulation TBS with 0.5% BSA and 0.02% sodium azide; pH 7.3
Gene DCXR
Gene Accession No. Q7Z4W1
Gene Alias 0610038K04Rik; 1810027P18Rik; carbonyl reductase; carbonyl reductase 2; carbonyl reductase II; DCR; Dcxr; diacetyl/L-xylulose reductase; dicarbonyl and L-xylulose reductase; dicarbonyl L-xylulose reductase; dicarbonyl/L-xylulose reductase; Glb; HCR2; HCRII; KI P34H; KID; KIDCR; Kidney dicarbonyl reductase; L-xylulose reductase; P34H; PNTSU; SDR20C1; Short chain dehydrogenase/reductase family 20C member 1; short chain dehydrogenase/reductase family 20C, member 1; sperm surface protein P34H; XR
Gene Symbols DCXR
Host Species Goat
Immunogen Synthetic peptide sequence (GSTLPVEGGFWAC) corresponding to the C-terminus amino acids of DCXR (aa 232-244).
Purification Method Ammonium sulfate precipitation
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 51181
Target Species Human
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.