Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ DCXR Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA582828
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA582828 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA582828 Supplier Invitrogen™ Supplier No. PA582828
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Immunogen sequence: IRVNAVNPTV VMTSMGQATW SDPHKAKTML NRIPLGKFAE VEHVVNAILF LLSDRSGMTT GSTLPVEG Highest antigen sequence indentity to the following orthologs: Rat - 82%, Mouse - 84%.

The protein encoded by this gene acts as a homotetramer to catalyze diacetyl reductase and L-xylulose reductase reactions. The encoded protein may play a role in the uronate cycle of glucose metabolism and in the cellular osmoregulation in the proximal renal tubules. Defects in this gene are a cause of pentosuria. Two transcript variants encoding different isoforms have been found for this gene.
TRUSTED_SUSTAINABILITY

Specifications

Antigen DCXR
Applications Immunohistochemistry (Paraffin), Western Blot
Classification Polyclonal
Concentration 0.2 mg/mL
Conjugate Unconjugated
Formulation PBS with 40% glycerol and 0.02% sodium azide; pH 7.2
Gene DCXR
Gene Accession No. Q7Z4W1
Gene Alias 0610038K04Rik; 1810027P18Rik; carbonyl reductase; carbonyl reductase 2; carbonyl reductase II; DCR; Dcxr; diacetyl/L-xylulose reductase; dicarbonyl and L-xylulose reductase; dicarbonyl L-xylulose reductase; dicarbonyl/L-xylulose reductase; Glb; HCR2; HCRII; KI P34H; KID; KIDCR; Kidney dicarbonyl reductase; L-xylulose reductase; P34H; PNTSU; SDR20C1; Short chain dehydrogenase/reductase family 20C member 1; short chain dehydrogenase/reductase family 20C, member 1; sperm surface protein P34H; XR
Gene Symbols DCXR
Host Species Rabbit
Immunogen Recombinant protein corresponding to Human DCXR. Recombinant protein control fragment (Product #RP-93411).
Purification Method Antigen affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 51181
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.