Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ DDA1 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA5100465
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA5100465 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA5100465 Supplier Invitrogen™ Supplier No. PA5100465
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Antibody detects endogenous levels of total DDA1.

Functions as a component of numerous distinct DCX (DDB1-CUL4-X-box) E3 ubiquitin-protein ligase complexes which mediate the ubiquitination and subsequent proteasomal degradation of target proteins (PubMed:17452440, PubMed:28437394, PubMed:28302793, PubMed:31686031, PubMed:31819272). In the DCX complexes, acts as a scaffolding subunit required to stabilize the complex (PubMed:31686031, PubMed:31819272). [UniProt].
TRUSTED_SUSTAINABILITY

Specifications

Antigen DDA1
Applications Western Blot
Classification Polyclonal
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation PBS with 50% glycerol and 0.02% sodium azide; pH 7.4
Gene DDA1
Gene Accession No. Q9BW61, Q9D9Z5
Gene Alias 1500034J01Rik; 1700095J19Rik; C19orf58; cross-immune reaction antigen PCIA1; Dda1; DET1 and DDB1 associated 1; DET1- and DDB1-associated protein 1; PCIA1; PCIA-1; Placenta cross-immune reaction antigen 1; R74921
Gene Symbols DDA1
Host Species Rabbit
Immunogen A synthesized peptide derived from human DDA1(Accession Q9BW61), corresponding to amino acid residues K65-T102.
Purification Method Affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 66498, 688813, 79016
Target Species Human, Mouse, Rat
Content And Storage -20°C
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.