Learn More
Invitrogen™ DDAH1 Recombinant Rabbit Monoclonal Antibody (1Q5F6)

Description
Immunogen sequence: IAIGSSESAQ KALKIMQQMS DHRYDKLTVP DDIAANCIYL NIPNKGHVLL HRTPEEYPES AKVYEKLKDH MLIPVSMSEL EKVDGLLTCC SVLINKKVDS.
Specifications
Specifications
| Antigen | DDAH1 |
| Applications | ELISA, Western Blot |
| Classification | Recombinant Monoclonal |
| Clone | 1Q5F6 |
| Concentration | 0.3 mg/mL |
| Conjugate | Unconjugated |
| Formulation | PBS with 0.05% BSA, 50% glycerol and 0.02% sodium azide; pH 7.3 |
| Gene | DDAH1 |
| Gene Accession No. | O08557, O94760 |
| Gene Alias | 2410006N07Rik; 2510015N06Rik; AI987801; AW050362; Ddah; Ddah1; DDAH-1; DDAHI; dimethylargininase-1; dimethylarginine dimethylaminohydrolase 1; epididymis secretory protein Li 16; FLJ21264; FLJ25539; HEL-S-16; N(G),N(G)-dimethylarginine dimethylaminohydrolase 1; NG, NG-dimethylarginine dimethylaminohydrolase; NG,NG dimethylarginine dimethylaminohydrolase; RP4-621F18.1 |
| Show More |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.