Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ DDAH1 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA587784
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA587784 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA587784 Supplier Invitrogen™ Supplier No. PA587784
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Immunogen sequence: MAGLGHPAAF GRATHAVVRA LPESLGQHAL RSAKGEEVDV ARAERQHQLY VGVLGSKLGL QVVELPADES LPDCVFVEDV AVVCEETALI TRPGAPSRRK EVDMMKEALE KLQLNIVEMK DENATLDGGD VLFTGREFFV GLSKRTNQRG AEILADTFKD YAVSTVPVAD GLHLKSFCSM AGPNLIAIGS SESAQKALKI MQQMSDHRYD KLTVPDDIAA NCIYLNIPNK GHVLLHRTPE EYPESAKVYE KLKDHMLIPV SMSELEKVDG LLTCCSVLIN KKVDS; Positive Samples: SW480, 22Rv1, HT-29, 293T, SGC-7901, Mouse brain, Mouse kidney, Rat brain.

This gene belongs to the dimethylarginine dimethylaminohydrolase (DDAH) gene family. The encoded enzyme plays a role in nitric oxide generation by regulating cellular concentrations of methylarginines, which in turn inhibit nitric oxide synthase activity.
TRUSTED_SUSTAINABILITY

Specifications

Antigen DDAH1
Applications ELISA, Western Blot, Immunocytochemistry
Classification Polyclonal
Concentration 3.47 mg/mL
Conjugate Unconjugated
Formulation PBS with 50% glycerol and 0.02% sodium azide; pH 7.3
Gene DDAH1
Gene Accession No. O08557, O94760, Q9CWS0
Gene Alias 2410006N07Rik; 2510015N06Rik; AI987801; AW050362; Ddah; Ddah1; DDAH-1; DDAHI; dimethylargininase-1; dimethylarginine dimethylaminohydrolase 1; epididymis secretory protein Li 16; FLJ21264; FLJ25539; HEL-S-16; N(G),N(G)-dimethylarginine dimethylaminohydrolase 1; NG, NG-dimethylarginine dimethylaminohydrolase; NG,NG dimethylarginine dimethylaminohydrolase; RP4-621F18.1
Gene Symbols DDAH1
Host Species Rabbit
Immunogen Recombinant fusion protein containing a sequence corresponding to amino acids 1-285 of human DDAH1 (NP_0362691).
Purification Method Affinity Chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 23576, 64157, 69219
Target Species Human, Mouse, Rat
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.