Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ DDAH1 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA517997
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA517997 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA517997 Supplier Invitrogen™ Supplier No. PA517997
Add to Cart
Add to Cart

Goat Polyclonal Antibody

This antibody is predicted to react with bovine based on sequence homology. This antibody is tested in Peptide ELISA: antibody detection limit dilution 1,000.

This gene belongs to the dimethylarginine dimethylaminohydrolase (DDAH) gene family. The encoded enzyme plays a role in nitric oxide generation by regulating cellular concentrations of methylarginines, which in turn inhibit nitric oxide synthase activity.
TRUSTED_SUSTAINABILITY

Specifications

Antigen DDAH1
Applications Flow Cytometry, Western Blot, Immunocytochemistry
Classification Polyclonal
Concentration 0.5 mg/mL
Conjugate Unconjugated
Formulation TBS with 0.5% BSA and 0.02% sodium azide; pH 7.3
Gene DDAH1
Gene Accession No. O94760
Gene Alias 2410006N07Rik; 2510015N06Rik; AI987801; AW050362; Ddah; Ddah1; DDAH-1; DDAHI; dimethylargininase-1; dimethylarginine dimethylaminohydrolase 1; epididymis secretory protein Li 16; FLJ21264; FLJ25539; HEL-S-16; N(G),N(G)-dimethylarginine dimethylaminohydrolase 1; NG, NG-dimethylarginine dimethylaminohydrolase; NG,NG dimethylarginine dimethylaminohydrolase; RP4-621F18.1
Gene Symbols DDAH1
Host Species Goat
Immunogen Synthetic peptide sequence (TCCSVLINKKVDS) corresponding to the C-terminus amino acids of DDAH1 (aa 273-285).
Purification Method Ammonium sulfate precipitation
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 23576
Target Species Human
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.