Learn More
DDAH1 Rabbit anti-Human, Polyclonal, Bethyl Laboratories
Rabbit Polyclonal Antibody
Supplier: Bethyl Laboratories, Inc A305391AT

Description
The recommended shelf life for this product is 1 year from date of receipt. Based on 100% sequence identity, this antibody is predicted to react with Bovine
This gene belongs to the dimethylarginine dimethylaminohydrolase (DDAH) gene family. The encoded enzyme plays a role in nitric oxide generation by regulating cellular concentrations of methylarginines, which in turn inhibit nitric oxide synthase activity.Specifications
DDAH1 | |
Polyclonal | |
Unconjugated | |
DDAH1 | |
DDAH1; dimethylarginine dimethylaminohydrolase 1; DDAH; DDAHI; NG, NG-dimethylarginine dimethylaminohydrolase; RP4-621F18.1; FLJ21264; FLJ25539 | |
Rabbit | |
Antigen affinity chromatography | |
RUO | |
23576 | |
4° C | |
Liquid |
Immunoprecipitation | |
1 mg/ml | |
phosphate, tris citrate with 0.09% sodium azide; pH 7-8 | |
O94760 | |
DDAH1 | |
Between 235 and 285 | |
10 μL | |
Primary | |
Human | |
Antibody | |
IgG |
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.