Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ DDAH2 Monoclonal Antibody (3E3)
GREENER_CHOICE

Catalog No. PIMA521504
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIMA521504 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIMA521504 Supplier Invitrogen™ Supplier No. MA521504
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

Peptide Sequence: MGTPGEGLGR CSHALIRGVP ESLASGEGAG AGLPALDLAK AQREHGVLGG KLRQRLGLRL LELPPEESLP LGPLLGDTAV IQGDTALITR PWSPARRPEV DGVRKALQDL GLRIVEIGDE NATLDGTDVL FTGREFFVGL SKWTNHRGAE IVADTFRDFA VSTVPVSGPS HLRGLCGMGG PRTVVAGSSD AAQKAVRAMA VLTDHPYASL TLPDDAAADC LFLRPGLPGV PPFLLHRGGG DLPNSQEALQ KLSDVTLVPV SCSELEKAGA GLSSLCLVLS TRPHS.

This gene belongs to the dimethylarginine dimethylaminohydrolase (DDAH) gene family. The encoded enzyme plays a role in nitric oxide generation by regulating cellular concentrations of methylarginines, which in turn inhibit nitric oxide synthase activity.
TRUSTED_SUSTAINABILITY

Specifications

Antigen DDAH2
Applications ELISA, Western Blot
Classification Monoclonal
Clone 3E3
Concentration 0.2-1.0 mg/mL
Conjugate Unconjugated
Formulation PBS with no preservative; pH 7.4
Gene Ddah2
Gene Accession No. O95865
Gene Alias 1110003M04Rik; AU019324; AW413173; Clone 7u; DADB-110M10.5; Ddah; DDAH2; DDAH-2; DDAHII; dimethylargininase-2; dimethylarginine dimethylaminohydrolase 2; dimethylarginine dimethylaminohydrolase II; epididymis secretory protein Li 277; G6a; HEL-S-277; N(G),N(G)-dimethylarginine dimethylaminohydrolase 2; NG30; OTTHUMP00000029307; OTTHUMP00000062666; OTTHUMP00000174406; Protein G6a; S-phase protein; testis tissue sperm-binding protein Li 54e
Gene Symbols Ddah2
Host Species Mouse
Immunogen DDAH2 (AAH01435, 1 a.a. ~ 285 a.a) full-length recombinant protein with GST tag. MW of the GST tag alone is 26 KDa.
Purification Method Affinity Chromatography
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 23564
Target Species Human
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Product Type Antibody
Form Liquid
Isotype IgG2b κ
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.