Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ DDAH2 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA5121004
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA5121004 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. PIPA5121004 Supplier Invitrogen™ Supplier No. PA5121004
Add to Cart
Add to Cart
This item is not returnable. View return policy

Rabbit Polyclonal Antibody

Positive test controls include: SW620, BT-474, MCF7, Mouse lung, Rat brain. The target is usually found in the following locations: Cytoplasm, Mitochondrion. Immunogen sequence: MGTPGEGLGR CSHALIRGVP ESLASGEGAG AGLPALDLAK AQREHGVLGG KLRQRLGLQL LELPPEESLP LGPLLGDTAV IQGDTALITR PWSPARRPEV DGVRKALQDL GLRIVEIGDE NATLDGTDVL FTGREFFVGL SKWTNHRGAE IVADTFRDFA VSTVPVSGPS HLRGLCGMGG PRTVVAGSSD AAQKAVRAMA VLTDHPYASL TLPDDAAADC LFLRPGLPGV PPFLLHRGGG DLPNSQEALQ KLSDVTLVPV SCSELEKAGA GLSSLCLVLS TRPHS.

This gene belongs to the dimethylarginine dimethylaminohydrolase (DDAH) gene family. The encoded enzyme plays a role in nitric oxide generation by regulating cellular concentrations of methylarginines, which in turn inhibit nitric oxide synthase activity.
TRUSTED_SUSTAINABILITY

Specifications

Antigen DDAH2
Applications ELISA, Immunohistochemistry (Paraffin), Western Blot, Immunocytochemistry
Classification Polyclonal
Concentration 0.68 mg/mL
Conjugate Unconjugated
Formulation PBS with 50% glycerol and 0.02% sodium azide; pH 7.3
Gene Ddah2
Gene Accession No. O95865, Q6MG60, Q99LD8
Gene Alias 1110003M04Rik; AU019324; AW413173; Clone 7u; DADB-110M10.5; Ddah; DDAH2; DDAH-2; DDAHII; dimethylargininase-2; dimethylarginine dimethylaminohydrolase 2; dimethylarginine dimethylaminohydrolase II; epididymis secretory protein Li 277; G6a; HEL-S-277; N(G),N(G)-dimethylarginine dimethylaminohydrolase 2; NG30; OTTHUMP00000029307; OTTHUMP00000062666; OTTHUMP00000174406; Protein G6a; S-phase protein; testis tissue sperm-binding protein Li 54e
Gene Symbols Ddah2
Host Species Rabbit
Immunogen Recombinant fusion protein containing a sequence corresponding to amino acids 1-285 of human DDAH2 (NP_039268.1).
Purification Method Affinity Chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 23564, 294239, 51793
Target Species Human, Mouse, Rat
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.