Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ DDAH2 Recombinant Rabbit Monoclonal Antibody (JB59-33)
GREENER_CHOICE

Catalog No. PIMA534681
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIMA534681 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIMA534681 Supplier Invitrogen™ Supplier No. MA534681
Add to Cart
Add to Cart

Rabbit Recombinant Monoclonal Antibody

Positive Control: Rat heart tissue lysate, mouse heart tissue lysate, mouse lung tissue lysate, human lung tissue lysate.

This gene belongs to the dimethylarginine dimethylaminohydrolase (DDAH) gene family. The encoded enzyme plays a role in nitric oxide generation by regulating cellular concentrations of methylarginines, which in turn inhibit nitric oxide synthase activity.
TRUSTED_SUSTAINABILITY

Specifications

Antigen DDAH2
Applications Immunoprecipitation, Western Blot
Classification Recombinant Monoclonal
Clone JB59-33
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation TBS with 0.05% BSA, 40% glycerol and 0.05% sodium azide; pH 7.4
Gene Ddah2
Gene Accession No. O95865
Gene Alias 1110003M04Rik; AU019324; AW413173; Clone 7u; DADB-110M10.5; Ddah; DDAH2; DDAH-2; DDAHII; dimethylargininase-2; dimethylarginine dimethylaminohydrolase 2; dimethylarginine dimethylaminohydrolase II; epididymis secretory protein Li 277; G6a; HEL-S-277; N(G),N(G)-dimethylarginine dimethylaminohydrolase 2; NG30; OTTHUMP00000029307; OTTHUMP00000062666; OTTHUMP00000174406; Protein G6a; S-phase protein; testis tissue sperm-binding protein Li 54e
Gene Symbols Ddah2
Host Species Rabbit
Immunogen Recombinant protein within Human DDAH2 aa 1-125.
Purification Method Protein A
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 23564
Target Species Human
Content And Storage -20°C, Avoid Freeze/Thaw Cycles, store in dark
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.