Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
DDB1 Antibody, Novus Biologicals™

Rabbit Polyclonal Antibody
Supplier: Novus Biologicals NBP133061
Description
DDB1 Polyclonal specifically detects DDB1 in Human, Mouse, Rat samples. It is validated for Western Blot, Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunoprecipitation, Immunohistochemistry-Paraffin.Specifications
DDB1 | |
Polyclonal | |
Unconjugated | |
1XPBS, 1% BSA, 20% Glycerol (pH7). with 0.025% Proclin 300 | |
damage-specific DNA binding protein 1 (127kD), damage-specific DNA binding protein 1, 127kDa, Damage-specific DNA-binding protein 1, DDB p127 subunit, DDBa, DNA damage-binding protein 1, DNA damage-binding protein a, HBV X-associated protein 1, UV-damaged DNA-binding factor, UV-damaged DNA-binding protein 1, UV-DDB 1, UV-DDB1, XAP1, XAP-1, Xeroderma pigmentosum group E-complementing protein, XPCe, XPE, XPE-BF, XPE-binding factor | |
Rabbit | |
127 kDa | |
0.1 mL | |
DNA Repair, Nucleotide Excision Repair | |
1642 | |
Human, Mouse, Rat | |
IgG |
Western Blot, Immunohistochemistry, Immunocytochemistry, Immunofluorescence, Immunoprecipitation, Immunohistochemistry (Paraffin) | |
0.81mg/mL | |
Western Blot 1:500-1:20000, Immunohistochemistry 1:100-1:1000, Immunocytochemistry/Immunofluorescence 1:100-1:1000, Immunoprecipitation 1:500-1:1000, Immunohistochemistry-Paraffin 1:100-1:1000, Immunoblotting 1:100 - 1:2000 | |
Q16531 | |
DDB1 | |
Recombinant protein encompassing a sequence within the C-terminus region of human DDB1. The exact sequence is proprietary. | |
Affinity Purified | |
RUO | |
Primary | |
Human and Mouse. Predicted cross-reactivity based on sequence identity: Chicken (98%), Xenopus laevis (96%), Bovine (100%). | |
Aliquot and store at -20C or -80C. Avoid freeze-thaw cycles. |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction