Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

DDB1 Mouse anti-Human, Mouse, Rat, Clone: 4F6C6, Proteintech
SDP

Catalog No. 501736453 Shop All Proteintech Group Inc Products
Encompass_Preferred
Change view
Click to view available options
Quantity:
150 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
50-173-6453 150 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 50-173-6453 Supplier Proteintech Group Inc Supplier No. 660101IG150UL
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

This antibody is specific to DDB1.

This gene encodes the large subunit of DNA damage-binding protein which is a heterodimer composed of a large and a small subunit. This protein functions in nucleotide-excision repair. Its defective activity causes the repair defect in the patients with xeroderma pigmentosum complementation group E (XPE). However, it remains for mutation analysis to demonstrate whether the defect in XPE patients is in this gene or the gene encoding the small subunit. In addition, Best vitelliform mascular dystrophy is mapped to the same region as this gene on 11q, but no sequence alternations of this gene are demonstrated in Best disease patients.

Specifications

Antigen DDB1
Applications Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin), Western Blot
Classification Monoclonal
Clone 4F6C6
Concentration 0.43 mg/mL
Conjugate Unconjugated
Formulation PBS with 50% glycerol and 0.02% sodium azide; pH 7.3
Gene DDB1
Gene Accession No. Q16531, Q3U1J4, Q9ESW0
Gene Alias DDB p127 subunit, DDB1, DDBA, DNA damage binding protein 1, DNA damage binding protein a, HBV X associated protein 1, UV damaged DNA binding factor, UV DDB 1, UV DDB1, XAP 1, XAP1, XPCE, XPE, XPE BF, XPE binding factor
Gene Symbols Ddb1
Host Species Mouse
Immunogen DDB1 Fusion Protein Ag17903
Purification Method Protein A
Quantity 150 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 13194, 1642, 64470
Target Species Human, Mouse, Rat
Content And Storage -20°C
Product Type Antibody
Form Liquid
Isotype IgG2b
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.