Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

DDB1 Polyclonal Antibody, Active Motif™
SDP

Catalog No. 501993167
Encompass
Change view
Click to view available options
Quantity:
10 μL
100 μL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
50-199-3167 10 μL
50-199-3166 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 50-199-3167 Supplier Active Motif Supplier No. 61128
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

The DNA damage-binding protein from HeLa cells is associated with polypeptides of relative mass 124,000 (DDB1) and 41,000 (DDB2) as determined by SDS-polyacrylamide gels. To test whether the DNA-repair defect in the subset of XPE patients that lack DNA damage-binding activity is caused by a defect in DDB, Keeney et al. (1994) injected purified human DDB protein into XPE cells.

Specifications

Antigen DDB1
Applications Western Blot
Classification Polyclonal
Concentration Lot-Specific
Conjugate Unconjugated
Formulation PBS with 30% glycerol and 0.035% sodium azide; pH 7.5
Gene DDB1
Gene Accession No. Q16531
Gene Alias 127kDa; AA408517; damage specific DNA binding protein 1; damaged-DNA recognition protein 1; damage-specific DNA binding protein 1, 127kDa; Damage-specific DNA-binding protein 1; damage-specific DNA-binding protein, DNA repair; DDB p127 subunit; DDB1; DDBa; DNA damage-binding protein 1; DNA damage-binding protein a; DNA repair protein; HBV X-associated protein 1; p127-Ddb1; UV-damaged DNA-binding factor; UV-damaged DNA-binding protein 1; UV-DDB 1; UV-DDB1; XAP1; XAP-1; xeroderma pigmento; Xeroderma pigmentosum group E-complementing protein; XPCe; XPE; XPE-BF; XPE-binding factor
Gene Symbols DDB1
Host Species Rabbit
Immunogen This DDB1 antibody was raised against a peptide from the C-terminus of human DDB1.
Purification Method Antigen affinity chromatography
Quantity 10 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 1642
Target Species Human
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.