Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

DDB1 Rabbit anti-Human, Mouse, Rat, Unconjugated, Clone: AFAH-4, Abnova™

Catalog No. 89339757 Shop All Abnova Corporation Products
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
89-339-757 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 89-339-757 Supplier Abnova Corporation Supplier No. MAB22019
Add to Cart
Add to Cart

Rabbit Monoclonal Antibody.

DDB1 Monoclonal antibody specifically detects DDB1 in Human, Mouse, Rat samples. It is validated for WB, IP, ICC, IF.

Specifications

Antigen DDB1
Applications Western Blot, Immunoprecipitation, Immunocytochemistry, Immunofluorescence
Classification Monoclonal
Clone AFAH-4
Conjugate Unconjugated
Description damage-specific DNA binding protein 1, 127kDa
Gene DDB1
Gene Alias DDBA, UV-DDB1, XAP1, XPCE, XPE, XPE-BF
Host Species Rabbit
Immunogen A synthetic peptide corresponding to human DDB1
Quantity 100 μg
Primary or Secondary Primary
Gene ID (Entrez) 1642
Target Species Human, Mouse, Rat
Content And Storage Store at 4°C. For long term storage store at -20°C. Aliquot to avoid repeated freezing and thawing.
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.