Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ DDHD2 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA554705
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA554705 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA554705 Supplier Invitrogen™ Supplier No. PA554705
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Immunogen sequence: GDKDNKYVPY SESFSQVLEE TYMLAVTLDE WKKKLESPNR EIIILHNPKL MVHYQPVAGS DDWGSTPTEQ GRPRTV Highest antigen sequence identity to the following orthologs: Mouse - 91%, Rat - 91%.

DDHD2, also known as KIAA0725p, is a member of the intracellular phospholipase A1 (PLA1) protein family which comprises a group of enzymes that hydrolyze the sn-1 ester bond of phospholipids, producing 2-acyl-lysophospholipids and fatty acids. DDHD2 prefers phosphatidic acid as substrate and has a role in efficient membrane trafficking from the Golgi apparatus to the plasma membrane. The gene of DDHD2 maps to chromosome 8p11.23, and encodes a 711-amino-acid protein with a calculated molecular mass of 81 kDa. It has been reported that DDHD2 immunoblot analysis of various tissue extracts revealed two bands of 90 and 85 kDa.
TRUSTED_SUSTAINABILITY

Specifications

Antigen DDHD2
Applications Immunohistochemistry (Paraffin)
Classification Polyclonal
Concentration 0.1 mg/mL
Conjugate Unconjugated
Formulation PBS with 40% glycerol and 0.02% sodium azide; pH 7.2
Gene DDHD2
Gene Accession No. O94830
Gene Alias 2010305K11Rik; DDHD domain containing 2; DDHD domain-containing protein 2; Ddhd2; intracellular phospholipase A1gamma; iPLA(1)gamma; KIAA0725; KIAA0725p; mKIAA0725; PA-PLA1 like; phospholipase DDHD2; SAM, WWE and DDHD domain-containing protein 1; Samwd1; sec23p-interacting protein p125-like phosphatidic acid-preferring phospholipase A1; SPG54
Gene Symbols DDHD2
Host Species Rabbit
Immunogen Recombinant protein corresponding to Human DDHD2. Recombinant protein control fragment (Product #RP-93745).
Purification Method Antigen affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 23259
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.