Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Invitrogen™ DDI1 Polyclonal Antibody
Rabbit Polyclonal Antibody
Supplier: Invitrogen™ PA527952
Description
Recommended positive controls: Molt-4. Predicted reactivity: Mouse (80%), Rat (82%), Rhesus Monkey (96%). Store product as a concentrated solution. Centrifuge briefly prior to opening the vial.
Ddi1 (DNA-damage inducible 1)appears to release substrate from an ubiquitination complex, making the substrate available for deubiquitination to facilitate proteasomal degradation. Ddi1 could be a potential selective target for lung cancer treatment. Ddi1 runs as a doublet of 54/56 kDa on acrylamide gels besides the calculated MW 44 kDa.
Specifications
DDI1 | |
Polyclonal | |
Unconjugated | |
DDI1 | |
1700011N24Rik; DDI1; DDI1, DNA-damage inducible 1, homolog 1; DDI1, DNA-damage inducible 1, homolog 1 (S. cerevisiae); DNA damage inducible 1 homolog 1; DNA-damage inducible 1 homolog 1; DNA-damage inducible 1 homolog 1 (S. cerevisiae); DNA-damage inducible protein 1; protein DDI1 homolog 1; RGD1559430 | |
Rabbit | |
Antigen affinity chromatography | |
RUO | |
414301 | |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
Liquid |
Immunohistochemistry (Paraffin), Western Blot, Immunocytochemistry | |
1 mg/mL | |
0.1M Tris glycine with 10% glycerol and 0.01% thimerosal; pH 7 | |
Q8WTU0 | |
DDI1 | |
Recombinant fragment corresponding to a region within amino acids 1 and 352 of Human DDI1. | |
100 μL | |
Primary | |
Human | |
Antibody | |
IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction