Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ DDI2 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA560282
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA560282 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA560282 Supplier Invitrogen™ Supplier No. PA560282
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Immunogen sequence: VDADFELHNF RALCELESGI PAAESQIVYA ERPLTDNHRS LASYGL Highest antigen sequence identity to the following orthologs: Mouse - 100%, Rat - 100%.

Aspartic protease that mediates the cleavage of NFE2L1/NRF1 at 'Leu-104', thereby promoting release of NFE2L1/NRF1 from the endoplasmic reticulum membrane. Ubiquitination of NFE2L1/NRF1 is a prerequisite for cleavage, suggesting that DDI2 specifically recognizes and binds ubiquitinated NFE2L1/NRF1.
TRUSTED_SUSTAINABILITY

Specifications

Antigen DDI2
Applications Immunohistochemistry (Paraffin), Immunocytochemistry
Classification Polyclonal
Concentration 0.2 mg/mL
Conjugate Unconjugated
Formulation PBS with 40% glycerol and 0.02% sodium azide; pH 7.2
Gene DDI2
Gene Accession No. Q5TDH0
Gene Alias 1110056G13Rik; 1700027M01Rik; 9130022E05Rik; AI604911; AU040698; DDI1, DNA-damage inducible 1, homolog 2; Ddi2; DNA damage inducible 1 homolog 2; DNA-damage inducible 1 homolog 2; DNA-damage inducible protein 2; DNA-damage inducible protein 2 (DDI2); protein DDI1 homolog 2; RP4-680D5.5
Gene Symbols DDI2
Host Species Rabbit
Immunogen Recombinant protein corresponding to Human DDI2. Recombinant protein control fragment (Product #RP-99834).
Purification Method Antigen affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 84301
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.