Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

DDI2 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA598500
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA598500 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA598500 Supplier Thermo Scientific Supplier No. PA598500
Add to Cart
Add to Cart

DDI2 Polyclonal Antibody

DDI2 Polyclonal Antibody for Western Blot, IHC (P), ELISA

TRUSTED_SUSTAINABILITY

Specifications

Antigen DDI2
Applications ELISA, Immunohistochemistry (Paraffin), Western Blot
Classification Polyclonal
Concentration 2 mg/mL
Conjugate Unconjugated
Formulation PBS with 50% glycerol and 0.03% ProClin 300; pH 7.4
Gene DDI2
Gene Accession No. Q5TDH0
Gene Alias 1110056G13Rik; 1700027M01Rik; 9130022E05Rik; AI604911; AU040698; DDI1, DNA-damage inducible 1, homolog 2; Ddi2; DNA damage inducible 1 homolog 2; DNA-damage inducible 1 homolog 2; DNA-damage inducible protein 2; DNA-damage inducible protein 2 (DDI2); protein DDI1 homolog 2; RP4-680D5.5
Gene Symbols DDI2
Host Species Rabbit
Immunogen Recombinant Human Protein DDI1 homolog 2 protein (1-211AA).
Purification Method Protein G
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 84301
Target Species Human
Content And Storage -20°C or -80°C if preferred
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.